Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha 4/CD49d Antibody (44H6), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. N1064903A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1064903A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1064903A70 Supplier Novus Biologicals Supplier No. NB10064903AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Integrin alpha 4/CD49d Monoclonal specifically detects Integrin alpha 4/CD49d in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) (Negative).

Specifications

Antigen Integrin alpha 4/CD49d
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) (Negative)
Classification Monoclonal
Clone 44H6
Conjugate Alexa Fluor 750
Dilution Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias 269C wild type, CD49 antigen-like family member D, CD49d, CD49d antigen, CD49Dantigen CD49D, alpha-4 subunit of VLA-4 receptor, IA4, integrin alpha 4, integrin alpha-4, integrin alpha-4 subunit, Integrin alpha-IV, integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor), MGC90518, very late activation protein 4 receptor, alpha 4 subunit, VLA-4 subunit alpha
Gene Symbols ITGA4
Host Species Mouse
Immunogen HOON pre-B leukemia cell line
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Cancer, Cellular Markers, Cytoskeleton Markers, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 3676.0
Test Specificity NB100-64903 recognizes CD49d, also known as Integrin alpha 4, which associates with either CD29 (integrin beta 1) or integrin beta 7. CD49d is a 150kD molecule, which may be cleaved to 80kD and 70kD fragments. It is expressed by lymphocytes, monocytes, eosinophils and thymocytes. VLA-4 mediates cell adhesion to VCAM-1 and fibronectin. 44H6 induces homotypic aggregation.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.