Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha 4/CD49d Antibody (44H6) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017661 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017661 0.025 mg
NB10064903 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017661 Supplier Novus Biologicals Supplier No. NB100649030.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Integrin alpha 4/CD49d Monoclonal specifically detects Integrin alpha 4/CD49d in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) (Negative).

Specifications

Antigen Integrin alpha 4/CD49d
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) (Negative)
Classification Monoclonal
Clone 44H6
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:40, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:50-1:100, Immunohistochemistry-Paraffin (Negative)
Formulation PBS (pH 7.2) with 0.09% Sodium Azide
Gene Accession No. P13612
Gene Alias 269C wild type, CD49 antigen-like family member D, CD49d, CD49d antigen, CD49Dantigen CD49D, alpha-4 subunit of VLA-4 receptor, IA4, integrin alpha 4, integrin alpha-4, integrin alpha-4 subunit, Integrin alpha-IV, integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor), MGC90518, very late activation protein 4 receptor, alpha 4 subunit, VLA-4 subunit alpha
Gene Symbols ITGA4
Host Species Mouse
Immunogen HOON pre-B leukemia cell line
Purification Method Protein A or G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Cytoskeleton Markers
Primary or Secondary Primary
Gene ID (Entrez) 3676
Test Specificity NB100-64903 recognizes CD49d, also known as Integrin alpha 4, which associates with either CD29 (integrin beta 1) or integrin beta 7. CD49d is a 150kD molecule, which may be cleaved to 80kD and 70kD fragments. It is expressed by lymphocytes, monocytes, eosinophils and thymocytes. VLA-4 mediates cell adhesion to VCAM-1 and fibronectin. 44H6 induces homotypic aggregation.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.