Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha 4/CD49d Antibody (7.2), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NBP250445AF Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250445AF 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250445AF Supplier Novus Biologicals Supplier No. NBP250445AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Integrin alpha 4/CD49d Monoclonal specifically detects Integrin alpha 4/CD49d in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen Integrin alpha 4/CD49d
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone 7.2
Conjugate Alexa Fluor 405
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 269C wild type, CD49 antigen-like family member D, CD49d, CD49d antigen, CD49Dantigen CD49D, alpha-4 subunit of VLA-4 receptor, IA4, integrin alpha 4, integrin alpha-4, integrin alpha-4 subunit, Integrin alpha-IV, integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor), MGC90518, very late activation protein 4 receptor, alpha 4 subunit, VLA-4 subunit alpha
Gene Symbols ITGA4
Host Species Mouse
Immunogen Human melanoma line DX3
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Cytoskeleton Markers
Primary or Secondary Primary
Gene ID (Entrez) 3676
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.