Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha 4/CD49d Antibody (HP2/1), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB12022858V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB12022858V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB12022858V Supplier Novus Biologicals Supplier No. NB12022858V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Integrin alpha 4/CD49d Monoclonal specifically detects Integrin alpha 4/CD49d in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen Integrin alpha 4/CD49d
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone HP2/1
Conjugate DyLight 405
Dilution Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 269C wild type, CD49 antigen-like family member D, CD49d, CD49d antigen, CD49Dantigen CD49D, alpha-4 subunit of VLA-4 receptor, IA4, integrin alpha 4, integrin alpha-4, integrin alpha-4 subunit, Integrin alpha-IV, integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor), MGC90518, very late activation protein 4 receptor, alpha 4 subunit, VLA-4 subunit alpha
Gene Symbols ITGA4
Host Species Mouse
Immunogen JM leukaemia line (Human).
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Cytoskeleton Markers, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 3676
Test Specificity Mouse anti Human CD49d monoclonal antibody, clone HP2/1 recognizes human CD49d also known as integrin alpha-4 or VLA-4 subunit alpha. CD49d is a ∼150kDa single pass type 1 transmembrane glycoprotein with seven FG-GAP repeats, characteristic of alpha integrins, in its extracellular domain. CD49d can be proteolytically cleaved to yield framents of 80 and 70kDa (Hemler et al. 1987). CD49d associates with either CD29 to form VLA-4 or with Integrin beta-7 to form The Peyer patches-specific homing receptor LPAM-1, involved in the lymphocyte migration and homing to gut-associated lymphoid tissue (Sackstein 2006) through its interaction with MadCam-1, preferentially expressed on Peyer's patch high endothelial venules and postcapillary venules in lamina propria (Briskin et al. 1997). Mouse anti human CD49d, clone HP2/1 binds to both intact and the 80kDa fragment of integrin alpha-4. CD49d is expressed on monocytes, T cells, B cells, thymocytes and Langerhans cells (de Graaf et al. 1995). Mouse anti Human CD49d, clone HP2/1 can be used in basic studies of VLA-4 mediated adhesion and its interaction with the VCAM-1 structure and has been demonstrated to inhibit cell binding to soluble VCAM-1 (Weller et al. 1991).
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.