Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha 4/CD49d Antibody (Natalizumab), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP27916A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP27916A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP27916A94 Supplier Novus Biologicals Supplier No. NBP275916AF594
Add to Cart
Add to Cart

Human Monoclonal Antibody

Integrin alpha 4/CD49d Monoclonal specifically detects Integrin alpha 4/CD49d in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Integrin alpha 4/CD49d
Applications Flow Cytometry
Classification Monoclonal
Clone Natalizumab
Conjugate Alexa Fluor 594
Dilution Flow Cytometry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias 269C wild type, CD49 antigen-like family member D, CD49d, CD49d antigen, CD49Dantigen CD49D, alpha-4 subunit of VLA-4 receptor, IA4, integrin alpha 4, integrin alpha-4, integrin alpha-4 subunit, Integrin alpha-IV, integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor), MGC90518, very late activation protein 4 receptor, alpha 4 subunit, VLA-4 subunit alpha
Gene Symbols ITGA4
Host Species Human
Immunogen The RAMOS cell line, a non-EBV releasing cell line, injected into mice.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3676
Test Specificity This antibody binds to the alpha 4 subunit of (alpha)4(beta)1 (also known as very late antigen 4 (VLA-4) or CD49d-CD29) and (alpha)4(beta)7 integrin
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG4 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.