Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha 5 beta 1 Antibody (BMC5), Novus Biologicals™
SDP

Catalog No. NB019463 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019463 0.025 mL
NBP229788 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019463 Supplier Novus Biologicals Supplier No. NBP2297880.025ML

Rat Monoclonal Antibody has been used in 2 publications

Integrin alpha 5 beta 1 Monoclonal specifically detects Integrin alpha 5 beta 1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, Functional, Immunohistochemistry-Paraffin (Negative).

Specifications

Antigen Integrin alpha 5 beta 1
Applications Functional Assay, Immunohistochemistry (Frozen), Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone BMC5
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry 10ul/ml, Immunohistochemistry 1:10-1:500, Immunoprecipitation 2 uL, Immunohistochemistry-Frozen 1:10-1:500, Functional 10ul/ml
Formulation Ascites with No Preservative
Gene Accession No. P08648
Gene Symbols ITGA5
Host Species Rat
Immunogen Mouse embryonal carcinoma P19 cells with myeloma P3X63Ag9.653 cells.
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3678
Test Specificity Antibody was raised against mouse integrin VLA-5 (alpha5beta1), a fibronectin receptor. The antibody was prepared by fusion of spleen cells from a Fisher rat immunized with mouse embryonal carcinoma P19 cells with myeloma P3X63Ag9.653 cells. Cell lines tested to express alpha5beta1, include: P19 cells, mouse melanoma B16F1 cells, mouse squamous carcoma SCC cells and thymocytes.
Target Species Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.