Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha 5 beta 1 Antibody (M200 (Volociximab)) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. N27590902M Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N27590902M 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N27590902M Supplier Novus Biologicals Supplier No. NBP2759090.2MG
Add to Cart
Add to Cart

Human Monoclonal Antibody

Integrin alpha 5 beta 1 Monoclonal specifically detects Integrin alpha 5 beta 1 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Block/Neutralize.

Specifications

Antigen Integrin alpha 5 beta 1
Applications Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Blocking Assay, Neutralization
Classification Monoclonal
Clone M200 (Volociximab)
Conjugate Unconjugated
Dilution Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Block/Neutralize
Formulation PBS with 0.02% Proclin 300
Gene Symbols ITGA5
Host Species Human
Immunogen Heterodimeric (alpha)5(beta)1Fc fusion protein
Quantity 0.2 mg
Primary or Secondary Primary
Test Specificity The antibody binds to human (alpha)5(beta)1 integrin with a Kd of 0.367 nM (determined by Biacore), an EC50 of 0.2 nM and an IC50 of 2.3 nM.
Target Species Human
Content And Storage Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
Isotype IgG4 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.