Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha 5 beta 1 Antibody (M200 (Volociximab)) - Chimeric - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP2526825 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2526825 0.025 mg
NBP25268002 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP2526825 Supplier Novus Biologicals Supplier No. NBP2526800.025MG
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody has been used in 5 publications

Integrin alpha 5 beta 1 Monoclonal specifically detects Integrin alpha 5 beta 1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunofluorescence, CyTOF-ready, Block/Neutralize.

Specifications

Antigen Integrin alpha 5 beta 1
Applications Flow Cytometry, ELISA, Immunohistochemistry, Blocking Assay, Immunofluorescence, CyTOF
Classification Monoclonal
Clone M200 (Volociximab)
Conjugate Unconjugated
Dilution Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Blocking/Neutralizing 50 - 200 molar excess, Immunofluorescence 1:10 - 1:500, CyTOF-ready
Formulation PBS with 0.02% Proclin 300. with 0.02% Proclin 300
Gene Symbols ITGA5
Host Species Rabbit
Immunogen Heterodimeric alpha 5 Beta 1 Fc fusion protein
Purification Method Protein A or G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity The antibody binds to human alpha 5 beta 1 integrin with a Kd of 0.367 nM (determined by Biacore), an EC50 of 0.2 nM and an IC50 of 2.3 nM.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.