Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha 5/CD49e Antibody (235112), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB367832 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB367832 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB367832 Supplier Novus Biologicals Supplier No. FAB1754E
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Integrin alpha 5/CD49e Monoclonal antibody specifically detects Integrin alpha 5/CD49e in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF

Specifications

Antigen Integrin alpha 5/CD49e
Applications Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 235112
Conjugate DyLight 405
Dilution Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD49 antigen-like family member E, CD49e, CD49e antigen, Fibronectin receptor subunit alpha, fibronectin receptor, alpha subunit, FNRAVLA5A, integrin alpha-5, Integrin alpha-F, integrin, alpha 5 (fibronectin receptor, alpha polypeptide), very late activation protein 5, alpha subunit, VLA-5
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Integrin alpha 5/CD49e, Phe45-Asn999 (Gln68Glu), Accession # P11688
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline CD Markers, Cell Biology, Cytoskeleton Markers, GPCR, Immunology, Signal Transduction, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 3678
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.