Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha 5/CD49e Antibody (CL6940), Novus Biologicals™

Mouse Monoclonal Antibody
$382.00 - $610.00
Specifications
Antigen | Integrin alpha 5/CD49e |
---|---|
Clone | CL6940 |
Classification | Monoclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
Integrin alpha 5/CD49e Monoclonal specifically detects Integrin alpha 5/CD49e in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
Integrin alpha 5/CD49e | |
Monoclonal | |
Purified | |
Human | |
CD49 antigen-like family member E, CD49e, CD49e antigen, Fibronectin receptor subunit alpha, fibronectin receptor, alpha subunit, FNRAVLA5A, integrin alpha-5, Integrin alpha-F, integrin, alpha 5 (fibronectin receptor, alpha polypeptide), very late activation protein 5, alpha subunit, VLA-5 | |
ITGA5 | |
IgG1 | |
Protein A purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
CL6940 | |
Unconjugated | |
Mouse | |
PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide | |
3678.0 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: DQPQKEEDLGPAVHHVYELINQGPSSISQGVLELSCPQALEGQQLLYVTRVTGLNCTTNHPINPKGLELDPEGSLHHQQKREAPSRSSASSGPQILKCPEAECFRLRCELGPLHQQESQSLQLHFRVWA | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title