Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha 5/CD49e Antibody (SNAKA52), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP250147FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250147FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250147FR Supplier Novus Biologicals Supplier No. NBP250147FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Integrin alpha 5/CD49e Monoclonal specifically detects Integrin alpha 5/CD49e in Human samples. It is validated for ELISA.

Specifications

Antigen Integrin alpha 5/CD49e
Applications ELISA
Classification Monoclonal
Clone SNAKA52
Conjugate DyLight 680
Dilution ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CD49 antigen-like family member E, CD49e, CD49e antigen, Fibronectin receptor subunit alpha, fibronectin receptor, alpha subunit, FNRAVLA5A, integrin alpha-5, Integrin alpha-F, integrin, alpha 5 (fibronectin receptor, alpha polypeptide), very late activation protein 5, alpha subunit, VLA-5
Gene Symbols ITGA5
Host Species Mouse
Immunogen alpha5 beta1 purified from human placenta.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3678
Test Specificity The binding of SNAKA52 is inhibited by anti-beta1 mAbs, indicating that its epitope is close to the interface between the alpha and beta subunits.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.