Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha 6 beta 1 Antibody (2548B), PE/Cy7, Novus Biologicals™
SDP

Catalog No. NB311105 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB311105 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB311105 Supplier Novus Biologicals Supplier No. FAB7809PECY7
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Integrin alpha 6 beta 1 Monoclonal antibody specifically detects Integrin alpha 6 beta 1 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen Integrin alpha 6 beta 1
Applications Flow Cytometry
Classification Monoclonal
Clone 2548B
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias CD49f, ITGA6B, VLA-6
Host Species Rabbit
Immunogen Chinese Hamster Ovary cell line, CHO-derived Human Integrin alpha 6 beta 1 heterodimer, Phe24-Ser1012(Integrin alpha 6) and Gln21-Asp728 (Integrin beta1), Accession # NP_000201
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3655
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.