Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha 6/CD49f Antibody (GoH3), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NBP143400W Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP143400W 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP143400W Supplier Novus Biologicals Supplier No. NBP143400AF405
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Integrin alpha 6/CD49f Monoclonal specifically detects Integrin alpha 6/CD49f in Human, Mouse, Bovine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen Integrin alpha 6/CD49f
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone GoH3
Conjugate Alexa Fluor 405
Dilution Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Gene Accession No. P23229
Gene Alias CD49 antigen-like family member F, CD49f, CD49f antigen, DKFZp686J01244, FLJ18737, integrin alpha-6, integrin alpha6B, integrin, alpha 6, ITGA6B, VLA-6
Gene Symbols ITGA6
Host Species Rat
Immunogen The immunogen for this antibody was CD49f.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3655
Test Specificity May also react with Bovine.
Target Species Human, Mouse, Bovine
Content And Storage Store at 4C in the dark.
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.