Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha 6/CD49f Antibody (GoH3), Biotin, Novus Biologicals™
SDP

Catalog No. NBP14363303 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP14363303 0.025 mg
NBP14363301 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP14363303 Supplier Novus Biologicals Supplier No. NBP1436330.025MG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Integrin alpha 6/CD49f Monoclonal specifically detects Integrin alpha 6/CD49f in Human, Mouse, Bovine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen Integrin alpha 6/CD49f
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone GoH3
Concentration 0.5 mg/mL
Conjugate Biotin
Dilution Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Formulation Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide
Gene Accession No. P23229
Gene Alias CD49 antigen-like family member F, CD49f, CD49f antigen, DKFZp686J01244, FLJ18737, integrin alpha-6, integrin alpha6B, integrin, alpha 6, ITGA6B, VLA-6
Gene Symbols ITGA6
Host Species Rat
Immunogen The immunogen for this antibody was CD49f.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3655
Test Specificity May also react with Bovine.
Target Species Human, Mouse, Bovine
Content And Storage Store at 4C. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.