Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha 6/CD49f Antibody (GoH3), Biotin, Novus Biologicals™
SDP

Rat Monoclonal Antibody

$268.00 - $622.00

Specifications

Antigen Integrin alpha 6/CD49f
Clone GoH3
Concentration 0.5 mg/mL
Dilution Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP14363303
SDP
View Documents
Novus Biologicals
NBP1436330.025MG
0.025 mg
Each for $268.00
Only null left
Add to Cart
 
NBP14363301
SDP
View Documents
Novus Biologicals
NBP1436330.1MG
0.1 mg
Each for $622.00
Only null left
Add to Cart
 
Description

Description

Integrin alpha 6/CD49f Monoclonal specifically detects Integrin alpha 6/CD49f in Human, Mouse, Bovine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications

Specifications

Integrin alpha 6/CD49f
0.5 mg/mL
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Biotin
RUO
Human, Mouse, Bovine
P23229
3655
The immunogen for this antibody was CD49f.
Primary
Store at 4C. Do not freeze.
GoH3
Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Monoclonal
Rat
Signal Transduction
Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide
CD49 antigen-like family member F, CD49f, CD49f antigen, DKFZp686J01244, FLJ18737, integrin alpha-6, integrin alpha6B, integrin, alpha 6, ITGA6B, VLA-6
ITGA6
IgG2a κ
Affinity Purified
May also react with Bovine.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.