Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha 6/CD49f Antibody (GoH3), PE/Atto594, Novus Biologicals™
SDP

Catalog No. NB314603 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB314603 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB314603 Supplier Novus Biologicals Supplier No. FAB13501PEATT594
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Integrin alpha 6/CD49f Monoclonal specifically detects Integrin alpha 6/CD49f in Bovine, Human, Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen Integrin alpha 6/CD49f
Applications Flow Cytometry
Classification Monoclonal
Clone GoH3
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS
Gene Alias CD49 antigen-like family member F, CD49f, CD49f antigen, DKFZp686J01244, FLJ18737, integrin alpha-6, integrin alpha6B, integrin, alpha 6, ITGA6B, VLA-6
Host Species Rat
Immunogen Mouse mammary tumor cells
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3655
Target Species Bovine, Human, Mouse
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.