Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha 6/CD49f Antibody (MP4F10), PE, Novus Biologicals™
SDP

Catalog No. NB297184 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297184 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB297184 Supplier Novus Biologicals Supplier No. FAB1350P
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Integrin alpha 6/CD49f Monoclonal specifically detects Integrin alpha 6/CD49f in Human samples. It is validated for CyTOF, Flow Cytometry, Immunohistochemistry, Immunoprecipitation.

Specifications

Antigen Integrin alpha 6/CD49f
Applications CyTOF, Flow Cytometry, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone MP4F10
Conjugate PE
Dilution Flow Cytometry, Immunohistochemistry, Immunoprecipitation, CyTOF-ready
Formulation PBS
Gene Alias CD49 antigen-like family member F, CD49f, CD49f antigen, DKFZp686J01244, FLJ18737, integrin alpha-6, integrin alpha6B, integrin, alpha 6, ITGA6B, VLA-6
Host Species Mouse
Immunogen SW1222 human colorectal carcinoma cell line
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3655
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.