Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha 6/CD49f Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP185747
Description
Integrin alpha 6/CD49f Polyclonal antibody specifically detects Integrin alpha 6/CD49f in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin).Specifications
Integrin alpha 6/CD49f | |
Polyclonal | |
Western Blot 0.04-0.4 ug/ml, Simple Western 1:50, Immunohistochemistry 1:200 - 1:500, Immunoprecipitation Reported in scientific literature (PMID: 23888051)., Immunohistochemistry-Paraffin 1:200 - 1:500 | |
P23229 | |
ITGA6 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:APYDDLGKVFIYHGSANGINTKPTQVLKGISPYFGYSIAGNMDLDRNSYPDVAVGSLSDSVTIFRSRPVINIQKTITVTPNRIDLRQKTACGAPSGICLQVKSCFEYTANPAGYNPSISIVGTLEAEKERRKSGLSS | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of human Integrin alpha 6/CD49f antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
CD49 antigen-like family member F, CD49f, CD49f antigen, DKFZp686J01244, FLJ18737, integrin alpha-6, integrin alpha6B, integrin, alpha 6, ITGA6B, VLA-6 | |
Rabbit | |
127 kDa | |
0.1 mL | |
Signal Transduction | |
3655 | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction