Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha 6/CD49f Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$684.50
Specifications
Antigen | Integrin alpha 6/CD49f |
---|---|
Dilution | Western Blot 0.04-0.4 ug/ml, Simple Western 1:50, Immunohistochemistry 1:200 - 1:500, Immunoprecipitation Reported in scientific literature (PMID: 23888051)., Immunohistochemistry-Paraffin 1:200 - 1:500 |
Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Integrin alpha 6/CD49f Polyclonal antibody specifically detects Integrin alpha 6/CD49f in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin).Specifications
Integrin alpha 6/CD49f | |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
P23229 | |
3655 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:APYDDLGKVFIYHGSANGINTKPTQVLKGISPYFGYSIAGNMDLDRNSYPDVAVGSLSDSVTIFRSRPVINIQKTITVTPNRIDLRQKTACGAPSGICLQVKSCFEYTANPAGYNPSISIVGTLEAEKERRKSGLSS | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
127 kDa |
Western Blot 0.04-0.4 ug/ml, Simple Western 1:50, Immunohistochemistry 1:200 - 1:500, Immunoprecipitation Reported in scientific literature (PMID: 23888051)., Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Polyclonal | |
Rabbit | |
Signal Transduction | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
CD49 antigen-like family member F, CD49f, CD49f antigen, DKFZp686J01244, FLJ18737, integrin alpha-6, integrin alpha6B, integrin, alpha 6, ITGA6B, VLA-6 | |
ITGA6 | |
IgG | |
Affinity Purified | |
Specificity of human Integrin alpha 6/CD49f antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title