Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha 6/CD49f Rabbit anti-Human, Clone: 4E4K6, Novus Biologicals™
SDP

Catalog No. NB166387 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166387 100 μg
NB166388 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB166387 Supplier Novus Biologicals Supplier No. NBP315646100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Integrin alpha 6/CD49f Monoclonal antibody specifically detects Integrin alpha 6/CD49f in Human samples. It is validated for Western Blot

Specifications

Antigen Integrin alpha 6/CD49f
Applications Western Blot
Classification Monoclonal
Clone 4E4K6
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias CD49 antigen-like family member F, CD49f, CD49f antigen, DKFZp686J01244, FLJ18737, integrin alpha-6, integrin alpha6B, integrin, alpha 6, ITGA6B, VLA-6
Host Species Rabbit
Immunogen A synthesized peptide derived from human Integrin alpha 6/CD49f (ITGA6/CD49f)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3655
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.