Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha 7 Antibody (334908) [DyLight 680], Novus Biologicals™
SDP

Catalog No. NB372136 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB372136 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB372136 Supplier Novus Biologicals Supplier No. FAB3518Y
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Integrin alpha 7 Monoclonal antibody specifically detects Integrin alpha 7 in Mouse samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen Integrin alpha 7
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 334908
Conjugate DyLight 680
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias FLJ25220, integrin alpha 7 chain, integrin alpha-7, integrin, alpha 7
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin alpha 7, Phe34-Pro1036, Accession # NP_032424
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3679
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.