Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha 8 Antibody (481709), HRP, Novus Biologicals™
SDP

Catalog No. NB347089 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB347089 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB347089 Supplier Novus Biologicals Supplier No. FAB6194H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Integrin alpha 8 Monoclonal antibody specifically detects Integrin alpha 8 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF

Specifications

Antigen Integrin alpha 8
Applications Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 481709
Conjugate HRP
Dilution Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation PBS
Gene Alias integrin alpha-8, integrin, alpha 8
Host Species Mouse
Immunogen E. coli-derived recombinant human Integrin alpha 8, Glu610-Leu800, Accession # P53708
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8516
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.