Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha D/CD11d Antibody (CA11.8H2), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. N1065292AP7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1065292AP7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1065292AP7 Supplier Novus Biologicals Supplier No. NB10065292APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Integrin alpha D/CD11d Monoclonal specifically detects Integrin alpha D/CD11d in Canine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation.

Specifications

Antigen Integrin alpha D/CD11d
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone CA11.8H2
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias ADB2, CD11 antigen-like family member D, CD11D, FLJ39841, integrin alpha-D, integrin, alpha D, leukointegrin alpha D
Gene Symbols ITGAD
Host Species Mouse
Immunogen Affinity purified CD11 antigen from canine spleen
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3681.0
Test Specificity NB100-65292 recognizes the canine CD11d cell surface antigen (the integrin ad sub-unit). The ad integrin is a newly recognized integrin chain that is expressed as a heterodimer with the b2 integrin (CD18). CD11d is expressed by a sub-population of CD8 positive T cells, macrophages and by gd T cells in the splenic red pulp.
Target Species Canine
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.