Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha E/CD103 Antibody (2G5.1), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10065271
Description
Integrin alpha E/CD103 Monoclonal specifically detects Integrin alpha E/CD103 in Human, Primate samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
Integrin alpha E/CD103 | |
Monoclonal | |
1.0 mg/mL | |
Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:50-1:100 | |
antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide, CD103, CD103 antigen, HML-1 antigen, HUMINAE, integrin alpha-E, Integrin alpha-IEL, integrin, alpha E (antigen CD103, human mucosal lymphocyte antigen 1; alphapolypeptide), MGC141996, Mucosal lymphocyte 1 antigen | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
NB100-65271 recognizes the human CD103 cell surface antigen, also known as the alphaE integrin. This reacts with various subsets of human intra-epithelial lymphocytes:- 40% of lamina propria T-cells; 30% of mesenteric lymphoblasts and some lymphocytes from other mucosa. It recognizes less than 0.1% bone marrow cells and less than 0.7% PBL. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2a |
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen) | |
2G5.1 | |
Unconjugated | |
P38570 | |
ITGAE | |
Human Intestinal lymphocyte | |
0.1 mg | |
Innate Immunity | |
3682 | |
Human, Primate | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction