Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha E/CD103 Antibody (Ber-ACT8), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NBP197564W Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP197564W 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP197564W Supplier Novus Biologicals Supplier No. NBP197564AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Integrin alpha E/CD103 Monoclonal specifically detects Integrin alpha E/CD103 in Human, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen Integrin alpha E/CD103
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone Ber-ACT8
Conjugate Alexa Fluor 405
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Gene Alias antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide, CD103, CD103 antigen, HML-1 antigen, HUMINAE, integrin alpha-E, Integrin alpha-IEL, integrin, alpha E (antigen CD103, human mucosal lymphocyte antigen 1; alphapolypeptide), MGC141996, Mucosal lymphocyte 1 antigen
Gene Symbols ITGAE
Host Species Mouse
Immunogen HTLV-1 induced human T cell line MAPS16
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3682
Test Specificity The mouse monoclonal antibody Ber-ACT8 recognizes an extracellular epitope of CD103 (alpha E integrin), a type I transmembrane glycoprotein primarily found on intestinal intraepithelial lymphocytes. HLDA V; WS Code A067
Target Species Human, Primate
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.