Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha E/CD103 Antibody (LF61), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB065272549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB065272549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB065272549 Supplier Novus Biologicals Supplier No. NB10065272JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IntegrinalphaE/CD103 Monoclonal specifically detects IntegrinalphaE/CD103 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation.

Specifications

Antigen IntegrinalphaE/CD103
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone LF61
Conjugate Janelia Fluor 549
Dilution Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide, CD103, CD103 antigen, HML-1 antigen, HUMINAE, integrin alpha-E, Integrin alpha-IEL, integrin, alpha E (antigen CD103, human mucosal lymphocyte antigen 1; alphapolypeptide), MGC141996, Mucosal lymphocyte 1 antigen
Gene Symbols ITGAE
Host Species Mouse
Immunogen Hairy cell leukemia cells
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3682
Test Specificity NB100-65272 recognizes the human CD103 cell surface antigen, a glycoprotein expressed by approximately 1% of peripheral blood lymphocytes, activated T lymphocytes and by hairy cell leukaemia cells. The antigen is also expressed by intraepithelial lymphocytes. It has recently been shown to be identical to the alpha E integrin.
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.