Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha E2/Dendritic Cells Antibody (OX-62), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB10065618R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10065618R 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10065618R Supplier Novus Biologicals Supplier No. NB10065618FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Integrin alpha E2/Dendritic Cells Monoclonal specifically detects Integrin alpha E2/Dendritic Cells in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot.

Specifications

Antigen Integrin alpha E2/Dendritic Cells
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone OX-62
Conjugate DyLight 680
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Gene Alias antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide, CD103, CD103 antigen, HML-1 antigen, HUMINAE, integrin alpha-E, Integrin alpha-IEL, integrin, alpha E (antigen CD103, human mucosal lymphocyte antigen 1; alphapolypeptide), MGC141996, Mucosal lymphocyte 1 antigen
Gene Symbols ITGAE
Host Species Mouse
Immunogen Density gradient enriched PVG rat thoracic duct dendritic cells.
Purification Method Protein G purified
Quantity 0.25 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3682
Test Specificity NB100-65618 recognizes the OX-62 antigen, shown to be rat alpha E2 integrin which is expressed by dendritic cells of dendritic epidermal T cells, intraepithelial lymphocytes in the small intestine and by cells of denritic morphology in lymphoid organs, at sites where gamma delta T cells are present.
Target Species Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.