Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha L/CD11a Antibody (345913), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB348289 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB348289 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB348289 Supplier Novus Biologicals Supplier No. FAB3595MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Integrin alpha L/CD11a Monoclonal specifically detects Integrin alpha L/CD11a in Human samples. It is validated for CyTOF, Flow Cytometry, Western Blot.

Specifications

Antigen Integrin alpha L/CD11a
Applications CyTOF, Flow Cytometry, Western Blot, CyTOF
Classification Monoclonal
Clone 345913
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Simple Western, Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD11 antigen-like family member A, CD11a antigen, CD11Aantigen CD11A (p180), lymphocyte function-associated antigen 1, alphapolypeptide, integrin alpha-L, integrin gene promoter, integrin, alpha L (antigen CD11A (p180), lymphocyte function-associated antigen1; alpha polypeptide), Leukocyte adhesion glycoprotein LFA-1 alpha chain, Leukocyte function-associated molecule 1 alpha chain, LFA-1, LFA-1 alpha, LFA1A, LFA-1A, lymphocyte function-associated antigen 1
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Integrin alpha L, Tyr26-Met1089 (Tyr660Ile), Accession # CAA68747
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cellular Markers, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Myeloid Cell Markers, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3683
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.