Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha V beta 3 Antibody (23C6), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB1002677X Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002677X 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002677X Supplier Novus Biologicals Supplier No. NB1002677AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Integrin alpha V beta 3 Monoclonal specifically detects Integrin alpha V beta 3 in Human samples. It is validated for CyTOF, Flow Cytometry, Immunohistochemistry, Immunoprecipitation.

Specifications

Antigen Integrin alpha V beta 3
Applications CyTOF, Flow Cytometry, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone 23C6
Conjugate Alexa Fluor 488
Dilution Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Gene Alias antigen identified by monoclonal L230, CD51, CD51 antigen, integrin alpha-V, integrin, alpha V (vitronectin receptor, alpha polypeptide, antigen CD51), MSK8, Vitronectin receptor subunit alpha, VNRADKFZp686A08142
Gene Symbols ITGAV
Host Species Mouse
Immunogen A cell suspension containing osteoclasts from osteoclastomas.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Angiogenesis, Cancer, Cytoskeleton Markers, Extracellular Matrix, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3685
Test Specificity Reacts with alpha V beta 3. 23C6 may be useful for bone resorption modulation, osteoclast identification, receptor purification, malignant melanoma identification and treatment. Human. Cross-reacts with rabbit and chicken, as reported in scientific literature.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.