Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha V beta 3 Antibody (23C6), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB002677549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002677549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002677549 Supplier Novus Biologicals Supplier No. NB1002677JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IntegrinalphaVbeta3 Monoclonal specifically detects IntegrinalphaVbeta3 in Human samples. It is validated for CyTOF, Flow Cytometry, Immunohistochemistry, Immunoprecipitation.

Specifications

Antigen IntegrinalphaVbeta3
Applications CyTOF, Flow Cytometry, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone 23C6
Conjugate Janelia Fluor 549
Dilution Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias antigen identified by monoclonal L230, CD51, CD51 antigen, integrin alpha-V, integrin, alpha V (vitronectin receptor, alpha polypeptide, antigen CD51), MSK8, Vitronectin receptor subunit alpha, VNRADKFZp686A08142
Gene Symbols ITGAV
Host Species Mouse
Immunogen A cell suspension containing osteoclasts from osteoclastomas.
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Adaptive Immunity, Angiogenesis, Cancer, Cytoskeleton Markers, Extracellular Matrix, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3685
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.