Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha V beta 3 Antibody (2549B), HRP, Novus Biologicals™
SDP

Catalog No. NB350093 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB350093 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB350093 Supplier Novus Biologicals Supplier No. FAB12192H
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Integrin alpha V beta 3 Monoclonal antibody specifically detects Integrin alpha V beta 3 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen Integrin alpha V beta 3
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 2549B
Conjugate HRP
Dilution Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias antigen identified by monoclonal L230, CD51, CD51 antigen, integrin alpha-V, integrin, alpha V (vitronectin receptor, alpha polypeptide, antigen CD51), MSK8, Vitronectin receptor subunit alpha, VNRADKFZp686A08142
Host Species Rabbit
Immunogen Chinese Hamster Ovary cell line CHO-derived human Integrin alpha V beta 3., Human Integrin alpha V (Phe31-Val992) and Human Integrin beta 3 (Gly27-Asp718), Accession # NP_002201 and AAA52589
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Angiogenesis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3685
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.