Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha V beta 5 Antibody (23C6) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB119751 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB119751 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB119751 Supplier Novus Biologicals Supplier No. NBP309128
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Integrin alpha V beta 5 Monoclonal specifically detects Integrin alpha V beta 5 in Human, Chicken samples. It is validated for Flow Cytometry, Immunoprecipitation, Immunohistochemistry-Frozen.

Specifications

Antigen Integrin alpha V beta 5
Applications Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 23C6
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry 2-6 ug/ml, Immunoprecipitation, Immunohistochemistry-Frozen
Formulation PBS
Gene Alias CD51, integrin subunit alpha V, ITGAV, MSK8, VNRA, VTNR
Host Species Mouse
Immunogen A cell suspension containing osteoclasts from osteoclastomas
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3685
Target Species Human, Chicken
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.