Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha V beta 5 Antibody (P5H9), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB364932 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB364932 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB364932 Supplier Novus Biologicals Supplier No. FAB2528Z
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Integrin alpha V beta 5 Monoclonal specifically detects Integrin alpha V beta 5 in Human samples. It is validated for Adhesion Blockade, CyTOF, Flow Cytometry, Immunocytochemistry, Immunoprecipitation.

Specifications

Antigen Integrin alpha V beta 5
Applications Adhesion Blockade, CyTOF, Flow Cytometry, Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone P5H9
Conjugate DyLight 755
Dilution Flow Cytometry, Immunoprecipitation, Adhesion Blockade, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD51, integrin subunit alpha V, ITGAV, MSK8, VNRA, VTNR
Host Species Mouse
Immunogen HT1080 human fibrosarcoma cell line
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3685
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.