Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha V beta 6 Antibody (Av beta6 62ow.17), HRP, Novus Biologicals™
SDP

Catalog No. NBP250450H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250450H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250450H Supplier Novus Biologicals Supplier No. NBP250450H
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Integrin alpha V beta 6 Monoclonal specifically detects Integrin alpha V beta 6 in Human, Mouse (Negative) samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Integrin alpha V + beta 6
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone Av beta6 62ow.17
Conjugate HRP
Dilution Western Blot, Immunohistochemistry
Formulation PBS with No Preservative
Host Species Rat
Immunogen Mouse fibroblasts retrovirally infected with cDNA to human beta6 (3T3 beta6.19). Cells expressed a mouse/human av beta6 chimera.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Cellular Signaling, Stem Cells
Primary or Secondary Primary
Test Specificity Does not recognize mouse av beta 6.
Target Species Human, Mouse (Negative)
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.