Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha V beta 6 Antibody (AvB6 53a.2), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP20449A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP20449A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP20449A94 Supplier Novus Biologicals Supplier No. NBP250449AF594
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Integrin alpha V beta 6 Monoclonal specifically detects Integrin alpha V beta 6 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Integrin alpha V beta 6
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone AvB6 53a.2
Conjugate Alexa Fluor 594
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CD51, integrin subunit alpha V, ITGAV, MSK8, VNRA, VTNR
Gene Symbols ITGAV
Host Species Rat
Immunogen Mouse fibroblasts retrovirally infected with cDNA to human Integrin alpha V beta 6 (3T3b6.19). Cells expressed a mouse/human Integrin alpha V beta 6 chimera.
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Cancer, Cardiovascular Biology, Cellular Signaling, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 3685
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.