Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha V beta 6 Antibody (AvB6 53a.2), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NBP250449AI Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250449AI 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250449AI Supplier Novus Biologicals Supplier No. NBP250449AF700
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Integrin alpha V beta 6 Monoclonal specifically detects Integrin alpha V beta 6 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Integrin alpha V + beta 6
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone AvB6 53a.2
Conjugate Alexa Fluor 700
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Host Species Rat
Immunogen Mouse fibroblasts retrovirally infected with cDNA to human Integrin alpha V beta 6 (3T3b6.19). Cells expressed a mouse/human Integrin alpha V beta 6 chimera.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Cellular Signaling, Stem Cells
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.