Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha V/CD51 Antibody (23C6), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB378528 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB378528 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB378528 Supplier Novus Biologicals Supplier No. NBP308850G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Integrin alpha V/CD51 Monoclonal antibody specifically detects Integrin alpha V/CD51 in Human, Chicken, Bovine (Negative), Rabbit (Negative) samples. It is validated for ELISA

Specifications

Antigen Integrin alpha V/CD51
Applications ELISA
Classification Monoclonal
Clone 23C6
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias antigen identified by monoclonal L230, CD51, CD51 antigen, integrin alpha-V, integrin, alpha V (vitronectin receptor, alpha polypeptide, antigen CD51), MSK8, Vitronectin receptor subunit alpha, VNRADKFZp686A08142
Host Species Mouse
Immunogen Osteoclasts from osteoclastomas
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Angiogenesis, Cancer, Cell Biology, Cellular Markers, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3685
Target Species Human, Chicken, Bovine (Negative), Rabbit (Negative)
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.