Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha V/CD51 Mouse anti-Human, Chicken, Clone: 23C6, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30752520UG
Description
Integrin alpha V/CD51 Monoclonal specifically detects Integrin alpha V/CD51 in Human, Chicken samples. It is validated for ELISA.Specifications
Integrin alpha V/CD51 | |
Monoclonal | |
0.2 mg/ml | |
ELISA | |
antigen identified by monoclonal L230, CD51, CD51 antigen, integrin alpha-V, integrin, alpha V (vitronectin receptor, alpha polypeptide, antigen CD51), MSK8, Vitronectin receptor subunit alpha, VNRADKFZp686A08142 | |
Osteoclasts from osteoclastomas | |
20 μg | |
Adaptive Immunity, Angiogenesis, Cancer, Cell Biology, Cellular Markers, Immunology, Signal Transduction | |
3685 | |
Store at 4°C. | |
IgG1 κ |
ELISA | |
23C6 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Chicken | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction