Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha V/CD51 Mouse anti-Human, Chicken, Clone: 23C6, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00
Specifications
| Antigen | Integrin alpha V/CD51 |
|---|---|
| Clone | 23C6 |
| Concentration | 0.2 mg/ml |
| Dilution | ELISA |
| Applications | ELISA |
Description
Integrin alpha V/CD51 Monoclonal specifically detects Integrin alpha V/CD51 in Human, Chicken samples. It is validated for ELISA.Specifications
| Integrin alpha V/CD51 | |
| 0.2 mg/ml | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Angiogenesis, Cancer, Cell Biology, Cellular Markers, Immunology, Signal Transduction | |
| 10 mM PBS with 0.05% BSA | |
| 3685 | |
| IgG1 κ | |
| Protein A or G purified |
| 23C6 | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Chicken | |
| antigen identified by monoclonal L230, CD51, CD51 antigen, integrin alpha-V, integrin, alpha V (vitronectin receptor, alpha polypeptide, antigen CD51), MSK8, Vitronectin receptor subunit alpha, VNRADKFZp686A08142 | |
| Osteoclasts from osteoclastomas | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title