Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin beta 1/CD29 Antibody (12G10), PerCP, Novus Biologicals™
SDP

Catalog No. NB10063255P Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10063255P 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10063255P Supplier Novus Biologicals Supplier No. NB10063255PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Integrin beta 1/CD29 Monoclonal specifically detects Integrin beta 1/CD29 in Human samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot.

Specifications

Antigen Integrin beta 1/CD29
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 12G10
Conjugate PerCP
Dilution Flow Cytometry
Gene Accession No. P05556
Gene Alias CD29, CD29 antigen, Fibronectin receptor subunit beta, FNRBVLAB, GPIIA, integrin beta-1, integrin VLA-4 beta subunit, integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includesMDF2, MSK12), MDF2, MSK12, very late activation protein, beta polypeptide, VLA-4 subunit beta, VLA-BETA
Gene Symbols ITGB1
Host Species Mouse
Immunogen Purified human beta1 integrin preparation from HT1080 fibrosarcoma cell extract.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cytokine Research, Cytoskeleton Markers, Embryonic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Neuronal Cell Markers, Neuronal Stem Cell Markers, Phospho Specific, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3688
Test Specificity NB100-63255 recognises the human beta1 integrin. This has been shown to bind to a ligand induced binding site, binding being increased in the presence of fibronectin ligand. This also enhances alpha 5 - beta 1 - fibronectin interactions.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.