Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin beta 1/CD29 Antibody (KMI6), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP143423A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP143423A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP143423A30 Supplier Novus Biologicals Supplier No. NBP143423AF350

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rat Monoclonal Antibody

Integrin beta 1/CD29 Monoclonal specifically detects Integrin beta 1/CD29 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen Integrin beta 1/CD29
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone KMI6
Conjugate Alexa Fluor 350
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CD29, CD29 antigen, Fibronectin receptor subunit beta, FNRBVLAB, GPIIA, integrin beta-1, integrin VLA-4 beta subunit, integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includesMDF2, MSK12), MDF2, MSK12, very late activation protein, beta polypeptide, VLA-4 subunit beta, VLA-BETA
Gene Symbols ITGB1
Host Species Rat
Immunogen The immunogen for this antibody was CD29.
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Cancer, Cytokine Research, Cytoskeleton Markers, Embryonic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Neuronal Cell Markers, Neuronal Stem Cell Markers, Phospho Specific, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3688.0
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.