Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin beta 1/CD29 Antibody (MEM-101A), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB500383UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500383UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500383UV Supplier Novus Biologicals Supplier No. NB500383UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Integrin beta 1/CD29 Monoclonal specifically detects Integrin beta 1/CD29 in Human, Porcine, Canine, Mouse (Negative) samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen Integrin beta 1/CD29
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone MEM-101A
Conjugate DyLight 350
Dilution Western Blot, Flow Cytometry
Gene Accession No. P05556
Gene Alias CD29, CD29 antigen, Fibronectin receptor subunit beta, FNRBVLAB, GPIIA, integrin beta-1, integrin VLA-4 beta subunit, integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includesMDF2, MSK12), MDF2, MSK12, very late activation protein, beta polypeptide, VLA-4 subunit beta, VLA-BETA
Gene Symbols ITGB1
Host Species Mouse
Immunogen Raji Burkitt's lymphoma cell line Raji (NM_002211.3).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cytokine Research, Cytoskeleton Markers, Embryonic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Neuronal Cell Markers, Neuronal Stem Cell Markers, Phospho Specific, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3688
Test Specificity The antibody MEM-101A reacts with CD29 (integrin beta1 chain), a 130 kDa single chain type I glycoprotein expressed as a heterodimer (non-covalently associated with the integrin alpha subunits 1-6). CD29 is broadly expressed on majority of hematopoietic and non-hematopoietic cells (leukocytes, platelets, fibroblasts, endothelial cells, epithelial cells and mast cells). HLDA VI; WS Code AS A048
Target Species Human, Pig, Canine, Mouse (Negative)
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.