Learn More
Description
Specifications
Specifications
| Antigen | Integrin beta 1D |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | 2B1 |
| Conjugate | Alexa Fluor 488 |
| Dilution | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen |
| Gene Alias | integrin beta-1, CD29, FNRB, GPIIA, integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includes MDF2, MSK12), MDF2, MSK12, VLAB, VLA-BETA |
| Gene Symbols | ITGB1 |
| Host Species | Mouse |
| Immunogen | Clone 2B1 is a mouse monoclonal IgG1, kappa antibody derived by fusion of SP2/0 mouse myeloma cells with spleen cells from a mouse immunized with a synthetic peptide corresponding to the C-terminal 24 amino acids of integrin beta1D including an appending N-terminal cysteine (CQENPIYKSPINNFKNPNYGRKAGL) coupled to keyhole limpet hemocyanin. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
