Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin beta 1D Antibody (2B1), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NBP97734549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP97734549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP97734549 Supplier Novus Biologicals Supplier No. NBP197734JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Integrin beta 1D Monoclonal specifically detects Integrin beta 1D in Human, Mouse, Porcine, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.

Specifications

Antigen Integrinbeta1D
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 2B1
Conjugate Janelia Fluor 549
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias integrin beta-1, CD29, FNRB, GPIIA, integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includes MDF2, MSK12), MDF2, MSK12, VLAB, VLA-BETA
Gene Symbols ITGB1
Host Species Mouse
Immunogen Clone 2B1 is a mouse monoclonal IgG1, kappa antibody derived by fusion of SP2/0 mouse myeloma cells with spleen cells from a mouse immunized with a synthetic peptide corresponding to the C-terminal 24 amino acids of integrin beta1D including an appending N-terminal cysteine (CQENPIYKSPINNFKNPNYGRKAGL) coupled to keyhole limpet hemocyanin.
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Cancer, Cytokine Research, Cytoskeleton Markers, Embryonic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Neuronal Cell Markers, Neuronal Stem Cell Markers, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3688
Test Specificity 2B1 recognizes specifically the cytoplasmic domain of integrin subunit beta 1D present in cardiac and skeletal muscle.
Target Species Canine, Human, Mouse, Pig
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.