Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin beta 1D Antibody (AT47E2), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB163274 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB163274 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB163274 Supplier Novus Biologicals Supplier No. NBP261161FR
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Integrin beta 1D Monoclonal antibody specifically detects Integrin beta 1D in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen Integrin beta 1D
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone AT47E2
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias integrin beta-1, CD29, FNRB, GPIIA, integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includes MDF2, MSK12), MDF2, MSK12, VLAB, VLA-BETA
Host Species Mouse
Immunogen Recombinant human Integrin beta 1D (21-461aa) purified from E. coli.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Mesenchymal Stem Cell Markers, Neuronal Cell Markers, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3688
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.