Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin beta 2/CD18 Antibody (212701), Biotin, Novus Biologicals™
SDP

Catalog No. NB304771 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB304771 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB304771 Supplier Novus Biologicals Supplier No. FAB1730B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Integrin beta 2/CD18 Monoclonal antibody specifically detects Integrin beta 2/CD18 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF

Specifications

Antigen Integrin beta 2/CD18
Applications Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 212701
Conjugate Biotin
Dilution Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation PBS
Gene Alias 959 beta subunit precursor)10, CD18 antigen, CD18beta 2, cell surface adhesion glycoprotein (LFA-1, CR3, integrin beta-2, integrin, beta 2 (antigen CD18 (p95), lymphocyte function-associated antigen 1,macrophage antigen 1 (mac-1) beta subunit), integrin, beta 2 (complement component 3 receptor 3 and 4 subunit), LAD, LCAMB, leukocyte cell adhesion molecule CD18, leukocyte-associated antigens CD18/11A, CD18/11B, CD18/11C, MF17, MFI7, P150
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Integrin beta 2/CD18, Gln23-Asn700, Accession # AAA59490
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cytokine Research, Immunology, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 3689
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.