Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin beta 2/CD18 Antibody (60.3), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB153487 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB153487 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB153487 Supplier Novus Biologicals Supplier No. NBP312030V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Integrin beta 2/CD18 Monoclonal antibody specifically detects Integrin beta 2/CD18 in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen Integrin beta 2/CD18
Applications Western Blot, ELISA
Classification Monoclonal
Clone 60.3
Conjugate DyLight 405
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias 959 beta subunit precursor)10, CD18 antigen, CD18beta 2, cell surface adhesion glycoprotein (LFA-1, CR3, integrin beta-2, integrin, beta 2 (antigen CD18 (p95), lymphocyte function-associated antigen 1;macrophage antigen 1 (mac-1) beta subunit), integrin, beta 2 (complement component 3 receptor 3 and 4 subunit), LAD, LCAMB, leukocyte cell adhesion molecule CD18, leukocyte-associated antigens CD18/11A, CD18/11B, CD18/11C, MF17, MFI7, P150
Host Species Mouse
Immunogen The original antibody was generated by immunizing BALB/c mice with alloantigen activated T cells.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cytokine Research, Immunology, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 3689
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.