Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin beta 2/CD18 Antibody (MEM-148), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB500480UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500480UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500480UV Supplier Novus Biologicals Supplier No. NB500480UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Integrin beta 2/CD18 Monoclonal specifically detects Integrin beta 2/CD18 in Human, Porcine (Negative) samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen Integrin beta 2/CD18
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone MEM-148
Conjugate DyLight 350
Dilution Western Blot, Flow Cytometry
Gene Accession No. P05107
Gene Alias 959 beta subunit precursor)10, CD18 antigen, CD18beta 2, cell surface adhesion glycoprotein (LFA-1, CR3, integrin beta-2, integrin, beta 2 (antigen CD18 (p95), lymphocyte function-associated antigen 1;macrophage antigen 1 (mac-1) beta subunit), integrin, beta 2 (complement component 3 receptor 3 and 4 subunit), LAD, LCAMB, leukocyte cell adhesion molecule CD18, leukocyte-associated antigens CD18/11A, CD18/11B, CD18/11C, MF17, MFI7, P150
Gene Symbols ITGB2
Host Species Mouse
Immunogen Human tissue cell preparation, Peripheral blood mononuclear cells (NM_000211.3).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cytokine Research, Immunology, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 3689
Test Specificity Recognizes the epitope exposed on free human CD18 chains unassociated with CD11 as well as on high-affinity state of LFA-1.
Target Species Human, Porcine (Negative)
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.