Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin beta 2/CD18 Antibody (YFC118.3), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB200610R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB200610R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB200610R Supplier Novus Biologicals Supplier No. NB200610R
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Integrin beta 2/CD18 Monoclonal specifically detects Integrin beta 2/CD18 in Human, Canine, Guinea Pig samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.

Specifications

Antigen Integrin beta 2/CD18
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone YFC118.3
Conjugate DyLight 550
Dilution Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias 959 beta subunit precursor)10, CD18 antigen, CD18beta 2, cell surface adhesion glycoprotein (LFA-1, CR3, integrin beta-2, integrin, beta 2 (antigen CD18 (p95), lymphocyte function-associated antigen 1;macrophage antigen 1 (mac-1) beta subunit), integrin, beta 2 (complement component 3 receptor 3 and 4 subunit), LAD, LCAMB, leukocyte cell adhesion molecule CD18, leukocyte-associated antigens CD18/11A, CD18/11B, CD18/11C, MF17, MFI7, P150
Gene Symbols ITGB2
Host Species Rat
Immunogen Human neutrophils
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3689
Test Specificity This recognises the human CD18 cell surface antigen, the integrin beta2 subunit. CD18 is expressed by the majority of leucocytes.
Target Species Human, Canine, Guinea Pig
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.