Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin beta 3/CD61 Antibody (909114), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB370846 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB370846 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB370846 Supplier Novus Biologicals Supplier No. FAB41182Y
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Integrin beta 3/CD61 Monoclonal specifically detects Integrin beta 3/CD61 in Mouse samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen Integrin beta 3/CD61
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 909114
Conjugate DyLight 680
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD61, CD61 antigen, GP3Aplatelet glycoprotein IIIa, GPIIIaintegrin beta-3, integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61), ITG B3, Platelet membrane glycoprotein IIIa
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin alpha 2b and Integrin beta 3 linked heterodimer, Leu32-Arg988 (Integrin alpha 2b) & Glu26-Asp717 (Integrin beta 3), Accession # NP_034705 (Integrin alpha 2b) & O54890 (Integrin beta 3)
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Immunology, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3690
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.